Merck
  • Adverse alterations in mitochondrial function contribute to type 2 diabetes mellitus-related endothelial dysfunction in humans.
Adverse alterations in mitochondrial function contribute to type 2 diabetes mellitus-related endothelial dysfunction in humans.
Arteriosclerosis, thrombosis, and vascular biology (2012-08-11)
Tinoy J Kizhakekuttu, Jingli Wang, Kodlipet Dharmashankar, Rong Ying, David D Gutterman, Joseph A Vita, Michael E Widlansky
ABSTRACT
Mitochondrial dysfunction plays a key pathophysiological role in type 2 diabetes mellitus (T2DM). Data delineating relationships between mitochondrial and endothelial dysfunction in humans with T2DM are lacking. In 122 human subjects (60 with T2DM, 62 without T2DM), we measured endothelial function by brachial artery ultrasound (flow mediated dilation) and digital pulse amplitude tonometery. Endothelial function in arterioles isolated from gluteal subcutaneous adipose was measured by videomicroscopy. In arterioles and mononuclear cells, we measured inner mitochondrial membrane potential (Δψ(m)), mitochondrial mass, and mitochondrial superoxide production using fluorophores. Endothelial function was impaired in T2DM subjects versus control subjects. Δψ(m) magnitude was larger and mitochondrial mass was lower in arterioles and mononuclear cells in T2DM. Mononuclear mitochondrial mass correlated with flow-mediated dilation and pulse amplitude tonometery (ρ=0.38 and 0.33, P=0.001 and 0.02, respectively), and mononuclear mitochondrial superoxide production inversely correlated with flow-mediated dilation (ρ=-0.58, P=0.03). Low doses of 2 different mitochondrial uncoupling agents (carbonyl cyanide m-chlorophenyl hydrazone and 2,4-dinitrophenol) that reduce Δψ(m) magnitude and a mitochondrial-targeted antioxidant (MitoTEMPOL) improved endothelial function and reduced mitochondrial superoxide levels in T2DM arterioles. Mitochondrial dysfunction may play a central role in the impairment of endothelial dysfunction in T2DM.
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
2,4-Dinitrophenol, moistened with water, ≥98.0%
Supelco
2,4-Dinitrophenol, PESTANAL®, analytical standard