Merck
  • Exocytosis of peptide functionalized gold nanoparticles in endothelial cells.
Exocytosis of peptide functionalized gold nanoparticles in endothelial cells.
Nanoscale (2012-06-30)
Dorota Bartczak, Simone Nitti, Timothy M Millar, Antonios G Kanaras
ABSTRACT
We present the exocytosis profile of two types of peptide-coated nanoparticles, which have similar charge and size but different functionality. While one kind of particles appears to progressively exocytose, the other one has a more complex profile, suggesting that some of the particles are re-uptaken by the cells. Both types of particles retain their colloidal stability after exocytosis.
Sigma-Aldrich
Hexaethylene glycol, 97%