Merck
  • PdCl2-catalyzed oxidative cycloisomerization of 3-cyclopropylideneprop-2-en-1-ones.
PdCl2-catalyzed oxidative cycloisomerization of 3-cyclopropylideneprop-2-en-1-ones.
Organic letters (2012-05-16)
Maozhong Miao, Jian Cao, Jingjing Zhang, Xian Huang, Luling Wu
ABSTRACT
A novel PdCl(2)-catalyzed oxidative cycloisomerization of 3-cyclopropylideneprop-2-en-1-ones, providing a facile synthesis of highly strained functionalized 2-alkylidenecyclobutanones via furan-fused cyclobutene intermediates, is reported. An interesting route to 2(3H)-furanones with a spiro-cyclopropane unit from the obtained 2-alkylidenecyclobutanones via a ring-contraction rearrangement reaction is also realized.
Sigma-Aldrich
Palladium(II) chloride solution, 5 wt. % in 10 wt. % HCl
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 60% Pd basis
Sigma-Aldrich
Palladium(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Palladium(II) chloride, 99.995%
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 59-60% palladium (Pd) basis
Sigma-Aldrich
Palladium(II) chloride, ≥99.9%