Merck
  • Synthesis of symmetric 1,4-diamino-2-butynes via a Cu(I)-catalyzed one-pot A3-coupling/decarboxylative coupling of a propiolic acid, an aldehyde, and an amine.
Synthesis of symmetric 1,4-diamino-2-butynes via a Cu(I)-catalyzed one-pot A3-coupling/decarboxylative coupling of a propiolic acid, an aldehyde, and an amine.
The Journal of organic chemistry (2012-05-09)
Huangdi Feng, Denis S Ermolat'ev, Gonghua Song, Erik V Van der Eycken
ABSTRACT
A novel microwave-assisted approach for the one-pot Cu(I)-catalyzed A(3)-coupling/decarboxylative coupling (PA(2)-coupling) of a propiolic acid, an aldehyde, and an amine, resulting in the formation of diversely substituted 1,4-diamino-2-butynes,is described. It is noteworthy that this new multicomponent coupling provides an efficient access to introduce alkyl and aryl group at the 1,4-position of the 1,4-diamino-2-butynes.
Sigma-Aldrich
Propiolic acid, 95%