Merck
  • Simultaneous analysis of strychnine and brucine and their major metabolites by liquid chromatography-electrospray ion trap mass spectrometry.
Simultaneous analysis of strychnine and brucine and their major metabolites by liquid chromatography-electrospray ion trap mass spectrometry.
Journal of analytical toxicology (2012-03-16)
Xueguo Chen, Yongquan Lai, Zongwei Cai
ABSTRACT
A liquid chromatography-electrospray ionization-ion trap mass spectrometry (LC-ESI-ITMS) method was developed for the simultaneous analysis of strychnine, brucine and their major metabolites. Strychnine and brucine were individually incubated with rat liver S9 fraction. The incubation samples were pooled together and analyzed with LC-ESI-ITMS in positive ion and full-scan detection mode. The calibration curves of strychnine and brucine in rat liver showed good linearity in ranges of 0.020 to 8.0 µg/mL for strychnine and 0.020 to 8.5 µg/mL for brucine. The limits of detections were both 0.008 µg/mL and the recoveries were 88.3 and 83.2% for strychnine and brucine, respectively. Two metabolites were identified as strychnine N-oxide and brucine N-oxide by comparing the molecular mass, retention time, full-scan mass spectra, tandem MS and MS(3) spectra with those of strychnine and brucine. The developed method provided high sensitivity and selectivity for the determination of poisonous alkaloids and their major metabolites and can be applied in the determination of samples in forensic and clinically toxicological cases.
Sigma-Aldrich
Brucine, anhydrous, 98%