Merck
  • Effect of the addition of silanated silica on the mechanical properties of microwave heat-cured acrylic resin.
Effect of the addition of silanated silica on the mechanical properties of microwave heat-cured acrylic resin.
Gerodontology (2012-01-10)
Lucas H da Silva, Sabrina A Feitosa, Marcia C Valera, Maria A M de Araujo, Rubens N Tango
ABSTRACT
The purpose of this study was to evaluate the flexural strength and Vickers hardness of a microwave energy heat-cured acrylic resin by adding different concentrations of silane surface-treated nanoparticle silica. Acrylic resin specimens with dimensions of 65 × 10 × 2.5 mm were formed and divided into five experimental groups (n = 10) according to the silica concentration added to the acrylic resin mass (weight %) prior to polymerisation : G1, without silica; G2, 0.1% silica; G3, 0.5% silica; G4, 1.0% silica; and G5, 5.0% silica. The specimens were submitted to a three-point flexural strength test and to the Vickers hardness test (HVN). The data obtained were statistically analysed by anova and the Tukey test (α = 0.05). Regarding flexural strength, G5 differed from the other experimental groups (G1, G2, G3 and G4) presenting the lowest mean, while G4 presented a significantly higher mean, with the exception of group G3. Regarding Vickers hardness, a decrease in values was observed, in which G1 presented the highest hardness compared with the other experimental groups. Incorporating surface-treated silica resulted in direct benefits in the flexural strength of the acrylic resin activated by microwave energy; however, similar results were not achieved for hardness.
Product Description
Supelco
Silanization solution I, ~5% (dimethyldichlorosilane in heptane), Selectophore
Sigma-Aldrich
Dichlorodimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Dichlorodimethylsilane, ≥99.5%
Sigma-Aldrich
Dichlorodimethylsilane, ≥98.5% (GC)