Merck
  • Direct perturbation theory in terms of energy derivatives: scalar-relativistic treatment up to sixth order.
Direct perturbation theory in terms of energy derivatives: scalar-relativistic treatment up to sixth order.
The Journal of chemical physics (2011-11-25)
Werner Schwalbach, Stella Stopkowicz, Lan Cheng, Jürgen Gauss
ABSTRACT
A formulation of sixth-order direct perturbation theory (DPT) to treat relativistic effects in quantum-chemical calculations is presented in the framework of derivative theory. Detailed expressions for DPT6 are given at the Hartree-Fock level in terms of the third derivative of the energy with respect to the relativistic perturbation parameter defined as λ(rel)=c(-2). They were implemented for the computation of scalar-relativistic energy corrections. The convergence of the scalar-relativistic DPT expansion is studied for energies and first-order properties such as dipole moment and electric-field gradient within the series of the hydrogen halides (HX, X = F, Cl, Br, I, and At). Comparison with spin-free Dirac-Coulomb calculations indicates that the DPT series exhibits a smooth and monotonic convergence. The rate of convergence, however, depends on the charge of the involved nuclei and significantly slows down for heavy-element compounds.
Sigma-Aldrich
Hydrobromic acid, 48 wt. % in H2O, ≥99.99%
Sigma-Aldrich
Hydrobromic acid, ACS reagent, 48%
Sigma-Aldrich
Hydrobromic acid, reagent grade, 48%