Merck
  • β₁-adrenergic receptor up-regulation induced by nadolol is mediated via signal transduction pathway coupled to α₁-adrenergic receptors.
β₁-adrenergic receptor up-regulation induced by nadolol is mediated via signal transduction pathway coupled to α₁-adrenergic receptors.
Brain research (2011-08-30)
Koji Mizuno, Kazuhiro Kurokawa, Masahiro Shibasaki, Seitaro Ohkuma
ABSTRACT
Although up-regulation of β-adrenergic receptors (β-ARs) occurs after long-term use of their antagonists in various tissues, the available data are little on mechanisms of β-AR up-regulation induced by their continuous blockade. The present study attempted to clarify mechanisms of β-AR up-regulation using mouse cerebral cortical neurons continuously exposed to nadolol (10 nM), a non-selective β-AR antagonist, for 24 h. Nadolol dose-dependently induced both subtypes of β-ARs, β₁- and β₂-ARs, which were not suppressed by protein A kinase inhibition with KT5720. On the other hand, blockade of α₁-ARs, which are immunohistochemically confirmed to be co-localized with β-ARs in the same neurons, significantly inhibited only β₁-AR up-regulation and the expression of β₂-ARs did not alter. In addition, phenylephrine, an agonist specific to α₁-ARs up-regulated β₁-ARs, but not β₂-ARs. Under the conditions with β-AR up-regulation, the level of phosphorylated protein kinase Cα (pPKCα) increased, which is significantly suppressed by prazosin, an α1-AR antagonist. Furthermore, nadolol decreased the degradation of mRNA of β₁-ARs, but not β₂-ARs. These results indicate that the nadolol-induced β₁-AR up-regulation is mediated via PKC-relating pathway via α₁-AR activation with stabilizing β₁-AR mRNA and that the increased expression of β₂-ARs is regulated by pathways different from those for β₁-AR expression.
Product Description
Supelco
Nadolol, analytical standard
Sigma-Aldrich
KT 5720, ≥98% (HPLC), powder