Merck
  • Outcomes of moderate-to-severe Pneumocystis pneumonia treated with adjunctive steroid in non-HIV-infected patients.
Outcomes of moderate-to-severe Pneumocystis pneumonia treated with adjunctive steroid in non-HIV-infected patients.
Antimicrobial agents and chemotherapy (2011-07-27)
Song Mi Moon, Tark Kim, Heungsup Sung, Mi-Na Kim, Sung-Han Kim, Sang-Ho Choi, Jin-Yong Jeong, Jun Hee Woo, Yang Soo Kim, Sang-Oh Lee
ABSTRACT
While it is well-known that adjunctive corticosteroid use improves the outcome of moderate-to-severe Pneumocystis jirovecii pneumonia (PcP) in patients with human immunodeficiency virus (HIV), there are limited data on its efficacy in non-HIV-infected patients with PcP. Patients undergoing fiber-optic bronchoscopy with bronchoalveolar lavage for suspected PcP from January 2007 through December 2010 were reviewed retrospectively. We compared demographics, clinical characteristics, and outcomes in 88 non-HIV-infected patients with moderate-to-severe PcP with (n = 59) and without (n = 29) adjunctive corticosteroid use. Outcomes of PcP were assessed by respiratory failure and 30-day and 90-day all-cause mortality. Survival curves were analyzed by the Kaplan-Meier method and estimated by the log rank test. All-cause mortality of moderate-to-severe PcP at 90 days was lower in the solid-organ transplant recipients than in all other patients (6/26 [23%] versus 34/62 [55%], respectively; P = 0.006), and mortality at 30 days was lower in patients with hematologic malignancies than in all other patients (4/26 [15%] versus 24/62 [39%], respectively; P = 0.03). The outcomes of PcP were not significantly different in moderate-to-severe PcP patients with and without adjunctive corticosteroid use, regardless of recent corticosteroid use. Survival analysis of PcP patients with and without corticosteroid use by the Kaplan-Meier method also did not reveal any difference (log rank test; P = 0.81). There again was no difference within the subgroup of PcP patients with solid-organ transplants. Adjunctive corticosteroid use may not improve the outcome of moderate-to-severe PcP in non-HIV-infected patients.
Product Description
Supelco
Sulfamethizole, VETRANAL®, analytical standard
Supelco
Sulfamethizole, analytical standard, ≥99% (HPLC)