Merck
  • Minimally invasive surface-enhanced Raman scattering detection with depth profiles based on a surface-enhanced Raman scattering-active acupuncture needle.
Minimally invasive surface-enhanced Raman scattering detection with depth profiles based on a surface-enhanced Raman scattering-active acupuncture needle.
Analytical chemistry (2011-07-07)
Jian Dong, Qingfeng Chen, Chunhui Rong, Danyang Li, Yanying Rao
ABSTRACT
To obtain depth profiles of surface-enhanced Raman scattering (SERS) information in living systems, a SERS-active needle was structured by acupuncture needles, gold nanoshells (GNSs), and polystyrene, which were used as carriers, SERS-active elements to be absorbed on the carriers, and coatings to protect the absorbed GNSs from being erased during insertion, respectively. The SERS-active needle is minimally invasive for entering and exiting the body. The interspaces between the GNSs became vessels to collect diffused fluids at different depths after a SERS-active needle was inserted into an agarose gel, and the SERS intensity profile on the SERS-active needle coincided with the concentration profile of Nile Blue A (NBA) in the gel. SERS detection in vitro avoided the signal attenuation in gels, and the SERS detection at different spots of the SERS-active needle provided a depth profile of the NBA molecule in the gel. In vivo experiments of NBA and 6-mercaptopurine confirmed that the SERS-active needle could collect fluids in living systems easily with minimal invasion and provide information about depth profiles of target molecules in tissues.
Sigma-Aldrich
Nile Blue A, certified by the Biological Stain Commission
Sigma-Aldrich
Nile Blue A, Dye content ≥75 %
Sigma-Aldrich
Nile Blue chloride, Dye content 85 %
Sigma-Aldrich
Nile Blue A perchlorate, Dye content 95 %