Merck
  • Degradation of 2,4,5-trichlorophenoxyacetic acid by a novel Electro-Fe(II)/Oxone process using iron sheet as the sacrificial anode.
Degradation of 2,4,5-trichlorophenoxyacetic acid by a novel Electro-Fe(II)/Oxone process using iron sheet as the sacrificial anode.
Water research (2011-05-10)
Y R Wang, W Chu
ABSTRACT
A novel electrochemically enhanced advanced oxidation process for the destruction of organic contaminants in aqueous solution is reported in this study. The process involves the use of an iron (Fe) sheet as sacrificial anode and a graphite bar as cathode. In the oxidation process, once an electric current is applied between the anode and the cathode, a predetermined amount of Oxone is added to the reactor. Ferrous ions generated from the sacrificed Fe anode mediate the generation of highly powerful radicals (SO(4)(•-)) through the decomposition of Oxone. The coupled process of Fe(II)/Oxone and electrochemical treatment (Electro-Fe(II)/Oxone) was evaluated in terms of 2,4,5-Trichlorophenoxyacetic acid degradation in aqueous solution. Various parameters were investigated to optimize the process, including applied current, electrolyte and Oxone concentration. In addition, low solution pH facilitates the system performance due to the dual effects of weak Fenton reagent generation and persulfate ions generation, whereas the system performance was inhibited at basic pH levels through non-radical self-dissociation of Oxone and the formation of ferric hydroxide precipitates. Furthermore, the active radicals involved in the Electro-Fe(II)/Oxone process were also identified. The Electro-Fe(II)/Oxone process demonstrates a very high 2,4,5-T degradation efficiency (over 90% decay within 10 min), which justifies the novel Electro-Fe(II)/Oxone a promising treatment process for herbicide removal in water.
Product Description
Supelco
2,4,5-T, PESTANAL®