Merck
  • Sequential anaerobic/aerobic biodegradation of chloroethenes--aspects of field application.
Sequential anaerobic/aerobic biodegradation of chloroethenes--aspects of field application.
Current opinion in biotechnology (2011-03-08)
Andreas Tiehm, Kathrin R Schmidt
ABSTRACT
Because of a range of different industrial activities, sites contaminated with chloroethenes are a world-wide problem. Chloroethenes can be biodegraded by reductive dechlorination under anaerobic conditions as well as by oxidation under aerobic conditions. The tendency of chloroethenes to undergo reductive dechlorination decreases with a decreasing number of chlorine substituents, whereas with less chlorine substituents chloroethenes more easily undergo oxidative degradation. There is currently a growing interest in aerobic metabolic degradation of chloroethenes, which demonstrates advantages compared to cometabolic degradation pathways. Sequential anaerobic/aerobic biodegradation can overcome the disadvantages of reductive dechlorination and leads to complete mineralization of the chlorinated pollutants. This approach shows promise for site remediation in natural settings and in engineered systems.
Sigma-Aldrich
Chloroethane solution, 2.0 M in tert-butyl methyl ether, anhydrous
Sigma-Aldrich
Chloroethane, ≥99.7%