- Catalytic conversion of carbohydrates into 5-hydroxymethylfurfural by germanium(IV) chloride in ionic liquids.
Catalytic conversion of carbohydrates into 5-hydroxymethylfurfural by germanium(IV) chloride in ionic liquids.
Direct conversion of carbohydrates into 5-hydroxymethylfurfural (HMF) catalyzed by germanium(IV) chloride in ionic liquids has been investigated in search of an efficient and environmentally friendly process. Monosaccharides D-fructose and D-glucose, disaccharides sucrose and maltose, and even the polysaccharide cellulose were successfully converted into HMF with good yields under mild conditions (yield up to 92ā% in 5ā min in the case of fructose). The structure of ionic liquids, catalyst loading, reaction temperature and water content had noticeable effects on this catalytic system. Addition of 5ā Ć molecular sieves during the dehydration of glucose resulted in an increase in HMF yield from 38.4ā% to 48.4ā%. A mechanism for glucose conversion to HMF catalyzed by germanium(IV) chloride was proposed according to Ā¹Ā³Cā NMR spectra obtained inā situ under different conditions using D-glucose-2-Ā¹Ā³C as the substrate.