Merck
  • Enhanced adhesion of dopamine methacrylamide elastomers via viscoelasticity tuning.
Enhanced adhesion of dopamine methacrylamide elastomers via viscoelasticity tuning.
Biomacromolecules (2010-12-25)
Hoyong Chung, Paul Glass, Jewel M Pothen, Metin Sitti, Newell R Washburn
ABSTRACT
We present a study on the effects of cross-linking on the adhesive properties of bio-inspired 3,4-dihydroxyphenylalanine (DOPA). DOPA has a unique catechol moiety found in adhesive proteins in marine organisms, such as mussels and polychaete, which results in strong adhesion in aquatic conditions. Incorporation of this functional group in synthetic polymers provides the basis for pressure-sensitive adhesives for use in a broad range of environments. A series of cross-linked DOPA-containing polymers were prepared by adding divinyl cross-linking agent ethylene glycol dimethacrylate (EGDMA) to monomer mixtures of dopamine methacrylamide (DMA) and 2-methoxyethyl acrylate (MEA). Samples were prepared using a solvent-free microwave-assisted polymerization reaction and compared to a similar series of cross-linked MEA materials. Cross-linking with EGDMA tunes the viscoelastic properties of the adhesive material and has the advantage of not reacting with the catechol group that is responsible for the excellent adhesive performance of this material. Adhesion strength was measured by uniaxial indentation tests, which indicated that 0.001 mol % of EGDMA-cross-linked copolymer showed the highest work of adhesion in dry conditions, but non-cross-linked DMA was the highest in wet conditions. The results suggest that there is an optimal cross-linking degree that displays the highest adhesion by balancing viscous and elastic behaviors of the polymer but this appears to depend on the conditions. This concentration of cross-linker is well below the theoretical percolation threshold, and we propose that subtle changes in polymer viscoelastic properties can result in significant improvements in adhesion of DOPA-based materials. The properties of lightly cross-linked poly(DMA-co-MEA) were investigated by measurement of the frequency dependence of the storage modulus (G') and loss modulus (G''). The frequency-dependence of G' and magnitude of G'' showed gradual decreases with the fraction of EGDMA. Loosely cross-linked DMA copolymers, containing 0% and 0.001 mol % of EGDMA-cross-linked copolymers, displayed rheological behavior appropriate for pressure-sensitive adhesives characterized by a higher G' at high frequencies and lower G' at low frequencies. Our results indicate that dimethacrylate cross-linking of DMA copolymers can be used to enhance the adhesive properties of this unique material.
Sigma-Aldrich
Methacrylamide, 98%