Merck
  • Solid lipid extrusion with small die diameters--electrostatic charging, taste masking and continuous production.
Solid lipid extrusion with small die diameters--electrostatic charging, taste masking and continuous production.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2010-10-14)
Rieke Witzleb, Venkata-Rangarao Kanikanti, Hans-Jürgen Hamann, Peter Kleinebudde
ABSTRACT
The aim of this work was to develop a continuous solid lipid extrusion process that includes post-process milling of the extrudates. Die diameters smaller than 0.5 mm should be used for taste masking of the bitter tasting anthelmintic praziquantel. During lipid extrusion with small die diameters, electrostatic charging of the extrudates occurred. This could be avoided by adding liquid polyethylene glycol (PEG) as antistatic agent. Further, extrusion with PEG as antistatic agent was possible with small diameter down to 0.2 mm and with up to 80% praziquantel load. Dissolution of praziquantel extrudates was shown to be faster with smaller extrudate diameter due to surface enlargement. Anyhow, different praziquantel extrudates with small diameter, drug load up to 70% and PEG content up to 20%, were proven to be sufficiently taste masked in a randomised palatability study with 40 cats. Within a scale-up experiment, lipid extrusion and milling of the extrudates in a centrifugal mill afterwards were conducted continuously. Extrudates from continuous and batchwise production revealed small differences in terms of size distribution and surface habit, but were similar in drug dissolution rate.
Sigma-Aldrich
Glyceryl tripalmitate, ≥85%
Sigma-Aldrich
Glyceryl tripalmitate, ≥99%