Merck
  • Opiate sparing effect of fixed combination of diclophenac and orphenadrine after unilateral total hip arthroplasty: A double-blind, randomized, placebo-controlled, multi-centre clinical trial.
Opiate sparing effect of fixed combination of diclophenac and orphenadrine after unilateral total hip arthroplasty: A double-blind, randomized, placebo-controlled, multi-centre clinical trial.
Wiener medizinische Wochenschrift (1946) (2010-10-05)
Hans Gombotz, Regina Lochner, Rudolf Sigl, Johann Blasl, Günther Herzer, Helmut Trimmel
ABSTRACT
Multimodal pain management combines analgesics to improve analgesia and reduce side effects. This study investigates the fixed combination of diclophenac and orphenadrin (Neodolpasse(®) Infusion Solution) in patients after unilateral total hip arthroplasty (THA). This prospective, randomized, double-blind, placebo-controlled, multi-centre clinical study enrolled 120 patients receiving patient-controlled analgesia (PCA). Isotonic saline was infused as placebo. The primary efficacy goal was defined as reduction of PCA analgesics used over the first 24 h post-surgery. The study used a three-stage group sequential test design with two interim analyses. Analgesia was monitored by visual analogue scale and verbal rating. Infusion of the Neodolpasse(®) Infusion Solution resulted in a significant reduction in the PCA analgesic requirements by approximately 30% (38.7 ± 21.3 mg vs. 55.9 ± 31.1 mg; p = 0.0004) while maintaining adequate analgesia and patient safety. This study demonstrates that Neodolpasse(®) Infusion Solution significantly reduces PCA analgesic requirements without compromising analgesic effectiveness and safety in THA patients.
Product Description
Orphenadrine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Orphenadrine hydrochloride, ≥98.0% (AT)
Supelco
Orphenadrine citrate salt, analytical standard