Merck
  • Grapevine cell early activation of specific responses to DIMEB, a resveratrol elicitor.
Grapevine cell early activation of specific responses to DIMEB, a resveratrol elicitor.
BMC genomics (2009-08-08)
Anita Zamboni, Pamela Gatto, Alessandro Cestaro, Stefania Pilati, Roberto Viola, Fulvio Mattivi, Claudio Moser, Riccardo Velasco
ABSTRACT
In response to pathogen attack, grapevine synthesizes phytoalexins belonging to the family of stilbenes. Grapevine cell cultures represent a good model system for studying the basic mechanisms of plant response to biotic and abiotic elicitors. Among these, modified beta-cyclodextrins seem to act as true elicitors inducing strong production of the stilbene resveratrol. The transcriptome changes of Vitis riparia x Vitis berlandieri grapevine cells in response to the modified beta-cyclodextrin, DIMEB, were analyzed 2 and 6 h after treatment using a suppression subtractive hybridization experiment and a microarray analysis respectively. At both time points, we identified a specific set of induced genes belonging to the general phenylpropanoid metabolism, including stilbenes and hydroxycinnamates, and to defence proteins such as PR proteins and chitinases. At 6 h we also observed a down-regulation of the genes involved in cell division and cell-wall loosening. We report the first large-scale study of the molecular effects of DIMEB, a resveratrol inducer, on grapevine cell cultures. This molecule seems to mimic a defence elicitor which enhances the physical barriers of the cell, stops cell division and induces phytoalexin synthesis.
Sigma-Aldrich
Heptakis(2,6-di-O-methyl)-β-cyclodextrin
Sigma-Aldrich
Heptakis(2,6-di-O-methyl)-β-cyclodextrin, ≥98.0% (TLC)