Merck
  • Efficacy of an injectable combination anthelmintic (nitroxynil+clorsulon+ivermectin) against early immature Fasciola hepatica compared to triclabendazole combination flukicides given orally or topically to cattle.
Efficacy of an injectable combination anthelmintic (nitroxynil+clorsulon+ivermectin) against early immature Fasciola hepatica compared to triclabendazole combination flukicides given orally or topically to cattle.
Veterinary parasitology (2009-04-21)
G W Hutchinson, K Dawson, C C Fitzgibbon, P J Martin
ABSTRACT
The objective was to compare the efficacy against artificially induced 2- and 4-week old early immature triclabendazole-susceptible liver flukes (Fasciola hepatica) of an injectable combination of nitroxynil, clorsulon and ivermectin with oral and pour-on combination formulations containing triclabendazole. Groups of yearling Angus or Angus cross cattle were confirmed fluke free before being artificially infected with 500 Sunny Corner strain triclabendazole-susceptible liver fluke metacercariae. Two or four weeks after infection, cattle were treated with the test combination Nitromec (10.2mg/kg nitroxynil, 2.0mg/kg clorsulon, 0.2mg/kg ivermectin), or oral Flukazole C+Se (triclabendazole/oxfendazole/Selenium), oral Fasimec C (triclabendazole/ivermectin) or Genesis Ultra Pour-On (triclabendazole/abamectin). At intervals cattle were weighed, faecal sampled for liver fluke egg counts and blood sampled for liver serum enzyme analysis. Cattle were slaughtered 14 weeks after infection for recovery of adult flukes; fluke egg counts and liver pathology assessment. All cattle increased in body weight by 0.4-0.8kg/day but there were no significant differences between control and treated groups or between the treatment groups. Geometric mean 14-week fluke egg counts and total fluke counts for all treatments, were significantly less (p<0.05) than the control group, except for the group treated with Genesis Ultra Pour-On, 2 weeks after infection. Nitromec treatment of 2-week old flukes was 83% and 95% effective as assessed by 14-week egg and fluke counts, respectively, compared to Flukazole C; 96% and 99%, Fasimec C; 70% and 46%, and Genesis Pour-On, which was ineffective, with egg and fluke count reductions of 0% and 8%, respectively. Against 4-week old flukes, Nitromec treatment was 88% and 99% effective when assessed by 14-week egg and fluke counts, respectively, with Flukazole C; 98% and 99%, Genesis Pour-On; 98% and 82% and Fasimec C; 91% and 61% effective, respectively. Group mean levels of the bile duct-associated enzyme gamma glutamyl transpeptidase (GGT) and the parenchymal associated enzymes, aspartate amino-transferase (AST) and glutamate dehydrogenase (GLDH) increased above the normal range 8 and 11 weeks after infection in the untreated control animals and the group treated 2 weeks after infection with Genesis Pour-On. The groups treated with Fasimec at 2 or 4 weeks after infection, also had elevated enzyme levels. The use of liver-associated enzyme assay is supported as supplementary indicators of fluke-induced pathology.
Product Description
Supelco
Nitroxinil, VETRANAL®, analytical standard
Supelco
Clorsulon, VETRANAL®, analytical standard
Supelco
Oxfendazole, VETRANAL®, analytical standard