Merck
  • Influence of aging on the acute depletion of reduced glutathione induced by electrophilic agents.
Influence of aging on the acute depletion of reduced glutathione induced by electrophilic agents.
Neurobiology of aging (1991-05-01)
G Benzi, O Pastoris, A Gorini, F Marzatico, R F Villa, D Curti
ABSTRACT
A severe age-dependent depletion of reduced glutathione (GSH) occurs in rat forebrain at 1-3 h from intraperitoneal injection of the electrophilic agents cyclohexene-1-one and cycloheptene-1-one. Chronic pretreatment with central dopamine agonists (i.e., ergot alkaloids; particularly, dihydroergocriptine) partially counteracts the GSH depletion induced in 15-month-old forebrains by the prooxidants tested. In contrast, chronic pretreatment with a vasodilator agent (i.e., papaverine) magnifies the GSH depletion.
Sigma-Aldrich
2-Cyclohepten-1-one, 80%, technical grade