Merck
  • Solution interactions between the uranyl cation [UO2(2+)] and histidine, N-acetyl-histidine, tyrosine, and N-acetyl-tyrosine.
Solution interactions between the uranyl cation [UO2(2+)] and histidine, N-acetyl-histidine, tyrosine, and N-acetyl-tyrosine.
Journal of inorganic biochemistry (2008-10-25)
Wei Xie, Ahmed Badawi, Huan Huang, J David Van Horn
ABSTRACT
Complexes of the uranyl cation [UO(2)(2+)] with histidine (His), N-acetyl-histidine (NAH), tyrosine (Tyr), and N-acetyl-tyrosine (NAT) were studied by UV-visible and NMR spectroscopy, and by potentiometric titration. Protonation constants for each ligand are reported, as are cumulative formation constants for uranyl-amino acid complexes. Coupling constant data (J(CH)) for uranyl-histidine complexes indicate that inner-sphere solution interactions between histidine and uranyl cation are solely at the carboxylate site. At 25 degrees C the major uranyl-histidine complex has a cumulative formation constant of logbeta(110)=8.53, and a proposed formula of [UO(2)HisH(2)(OH)(2)](+); the stepwise formation constant, logK(UL), is estimated to be 5.6 ( approximately 8.53-(-6.1)-(-6.1)-15.15). Outer-sphere interactions, H-bonding or electrostatic interactions, are proposed as contributing a significant portion of the stability to the ternary uranyl-hydroxo-amino acid complexes. The temperature dependent protonation constants of histidine and formation constants between uranyl cation and histidine are reported from 10 to 35 degrees C; at 25 degrees C, DeltaG=-43.3 kJ/mol.
Product Description
Supelco
N-Acetyl-L-tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
N-Acetyl-L-tyrosine