Merck
  • Ring expansion of lactones and lactams via propiolate 1-carbon intercalation.
Ring expansion of lactones and lactams via propiolate 1-carbon intercalation.
Organic letters (2008-08-19)
Tina N Grant, Chantel L Benson, F G West
ABSTRACT
Readily available five- and six-membered lactones and N-sulfonyllactams undergo efficient addition of t-butyl propiolate, and the resulting adducts undergo cycloisomerization to six- and seven-membered cyclic ethers or amines in the presence of pyridinium acetate. The ring expansion process occurs in generally good yields and is proposed to involve a nucleophilic catalysis mechanism.
Sigma-Aldrich
Propiolic acid, 95%