Merck
  • Presynaptic release probability and readily releasable pool size are regulated by two independent mechanisms during posttetanic potentiation at the calyx of Held synapse.
Presynaptic release probability and readily releasable pool size are regulated by two independent mechanisms during posttetanic potentiation at the calyx of Held synapse.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2008-08-08)
Jae Sung Lee, Myoung-Hwan Kim, Won-Kyung Ho, Suk-Ho Lee
ABSTRACT
At the immature calyx of Held, the fast decay phase of a Ca(2+) transient induced by tetanic stimulation (TS) was followed by a period of elevated [Ca(2+)](i) for tens of seconds, referred to as posttetanic residual calcium (Ca(res)). We investigated the source of Ca(res) and its contribution to posttetanic potentiation (PTP). After TS (100 Hz for 4 s), posttetanic Ca(res) at the calyx of Held was largely abolished by tetraphenylphosphonium (TPP(+)) or Ru360, which inhibit mitochondrial Na(+)-dependent Ca(2+) efflux and Ca(2+) uniporter, respectively. Whereas the control PTP lasted longer than Ca(res), inhibition of Ca(res) by TPP(+) resulted in preferential suppression of the early phase of PTP, the decay time course of which well matched with that of Ca(res). TS induced significant increases in release probability (P(r)) and the size of the readily releasable pool (RRP), which were estimated from plots of cumulative EPSC amplitudes. TPP(+) or Ru360 suppressed the posttetanic increase in P(r), whereas it had little effect on the increase in RRP size. Moreover, the posttetanic increase in P(r), but not in RRP size, showed a linear correlation with the amount of Ca(res). In contrast, myosin light chain kinase (MLCK) inhibitors and blebbistatin reduced the posttetanic increase in RRP size with no effect on the increase in P(r). Application of TPP(+) in the presence of MLCK inhibitor peptide caused further suppression of PTP. These findings suggest that Ca(res) released from mitochondria and activation of MLCK are primarily responsible for the increase in P(r) and that in the RRP size, respectively.
Sigma-Aldrich
Tetraphenylphosphonium chloride, 98%
Sigma-Aldrich
Tetraphenylphosphonium bromide, 97%