Merck
Synthesis and spectroscopic studies of new Schiff bases.
Molecules (Basel, Switzerland) (2007-10-27)
Hamid Latif Siddiqui, Amjid Iqbal, Saeed Ahmad, W Weaver
ABSTRACT
Five novel Schiff bases have been prepared from o-formylphenoxyacetic acid and a series of aminothiazoles to form a number of potentially biologically active compounds. The structures of these Schiff bases have been characterized using IR and (1)H-and (13)C-NMR spectroscopy.
Sigma-Aldrich
2-Amino-1,3,4-thiadiazole, 97%