Merck
  • Efficient synthesis of taurine and structurally diverse substituted taurines from aziridines.
Efficient synthesis of taurine and structurally diverse substituted taurines from aziridines.
The Journal of organic chemistry (2007-05-15)
Libo Hu, Hui Zhu, Da-Ming Du, Jiaxi Xu
ABSTRACT
Taurine and substituted taurines were synthesized efficiently from aziridines via ring-opening reaction with thioacetic acid, oxidation with performic acid, and hydrolysis in hydrochloric acid. The current method shows more benefit in purification and efficiency in the preparation of taurine and structurally diverse 2-substituted, 2,2-disubstituted, and 1,2-, 2,2-, and 2,N-alkylene taurines.
Sigma-Aldrich
Thioacetic acid, 96%
Sigma-Aldrich
Thioacetic acid, 96%
Sigma-Aldrich
Potassium thioacetate, 98%