Merck
  • 2-methoxycinnamaldehyde reduces IL-1beta-induced prostaglandin production in rat cerebral endothelial cells.
2-methoxycinnamaldehyde reduces IL-1beta-induced prostaglandin production in rat cerebral endothelial cells.
Biological & pharmaceutical bulletin (2006-11-02)
Jian-You Guo, Hai-Ru Huo, Yuan-Xiao Yang, Cang-Hai Li, Hong-Bin Liu, Bao-Sheng Zhao, Lan-Fang Li, Yue-Ying Ma, Shu-Ying Guo, Ting-Liang Jiang
ABSTRACT
Prostaglandin E2 (PGE2) works as a common final mediator of the febrile. Guizhi-Tang, one of the most famous traditional Chinese medical formula used to treat influenza, common cold and other pyretic conditions, was previously reported to reduce the production of PGE 2 in rats. 2-Methoxycinnamaldehyde is a principle compound isolated from Guizhi-Tang. The aim of the present study was to investigate the effects of 2-methoxycinnamaldehyde on PGE2 production of rat cerebral endothelial cells (CECs). 2-Methoxycinnamaldehyde dose-dependently inhibited interleukin (IL)-1beta-induced PGE2 production in CECs with IC50 values of 174 microM. IL-1beta stimulation increased the protein, activity and mRNA expression of cyclooxygenase (COX)-2 but not COX-1. 2-Methoxycinnamaldehyde reduced IL-1beta-induced protein and activity of COX-2, but did not influence the COX-2 mRNA expression. Our results show that prostaglandin production in CECs during stimulated conditions is sensitive to inhibition by 2-methoxycinnamaldehyde and suggest that 2-methoxycinnamaldehyde may reduce COX-2 protein level and activity but not COX-2 mRNA.
Sigma-Aldrich
2-Methoxycinnamaldehyde, natural, 98%, FG
Sigma-Aldrich
2-Methoxycinnamaldehyde, predominantly trans, 96%