Merck
  • Electron affinities of polycyclic aromatic hydrocarbons by means of B3LYP/6-31+G* calculations.
Electron affinities of polycyclic aromatic hydrocarbons by means of B3LYP/6-31+G* calculations.
The journal of physical chemistry. A (2006-05-19)
Alberto Modelli, Laura Mussoni, Daniele Fabbri
ABSTRACT
The gas-phase experimental adiabatic electron affinities (AEAs) of the polycyclic aromatic hydrocarbons (PAHs) anthracene, tetracene, pentacene, chrysene, pyrene, benzo[a]pyrene, benzo[e]pyrene, and fluoranthene are well reproduced using the hybrid density functional method B3LYP with the 6-31+G* basis set, indicating that the smallest addition of diffuse functions to the basis set is suitable for a correct description of the stable PAH anion states. The calculated AEAs also give a very good linear correlation with available reduction potentials measured in solution. The AEAs (not experimentally available) of the isomeric benzo[ghi]fluoranthene and cyclopenta[cd]pyrene, commonly found in the environment, are predicted to be 0.817 and 1.108 eV, respectively, confirming the enhancement of the electron-acceptor properties associated with fusion of a peripheral cyclopenta ring. The calculated localization properties of the lowest unoccupied MO of cyclopenta[cd]pyrene, together with its relatively high electron affinity, account for a high reactivity at the ethene double bond of this PAH in reductive processes.
Product Description
Supelco
Benzo[k]fluoranthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Benzo[k]fluoranthene, BCR®, certified reference material
Sigma-Aldrich
Benzo[k]fluoranthene, for fluorescence, ≥99%