Merck
  • DBU assisted expeditious synthesis of glycosyl dienes via glycosylated beta-hydroxy esters.
DBU assisted expeditious synthesis of glycosyl dienes via glycosylated beta-hydroxy esters.
Carbohydrate research (2006-05-16)
Shyam S Verma, Namrata Dwivedi, Biswajit K Singh, Rama P Tripathi
ABSTRACT
DBU catalyzed condensation of 3-O-benzyl(methyl)-5,6-dideoxy-1,2-O-isopropylidene-beta-L-threo-hept-4-enofuranuronates with different aldehydes produces the corresponding 3-O-benzyl(methyl)-6-carbethoxy-5,6-dideoxy-1,2-O-isopropylidene-7-phenyl-beta-L-threo-hept-4-enofuranoses. The latter on treatment with methanesulfonyl chloride followed by DBU catalyzed E2 reaction of the methanesulfonyloxy intermediates gave the respective 3-O-benzyl(methyl)-6-carbethoxy-5,6,7-trideoxy-1,2-O-isopropylidene-7-phenyl-beta-L-threo-hept-4,6-dienofuranose in moderate to good yields.
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98%
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene, puriss., ≥99.0% (GC)
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene solution, 1 M in THF