Merck
  • Biosynthesis of medium-chain-length poly(hydroxyalkanoates) from soy molasses.
Biosynthesis of medium-chain-length poly(hydroxyalkanoates) from soy molasses.
Biotechnology letters (2006-02-21)
Daniel K Y Solaiman, Richard D Ashby, Arland T Hotchkiss, Thomas A Foglia
ABSTRACT
Pseudomonas corrugata was selected from a screening process for the bioconversion of inexpensive soy molasses into medium-chain-length poly(hydroxyalkanoates) (mcl-PHA). We obtained yields of 1.5 g cell dry weight (CDW)/l culture with growth medium supplemented with 2% (w/v) soy molasses, and of an average of 3.4 g CDW/l with 5% (w/v) soy molasses. Crude PHAs were obtained at 5-17% of CDW. The most prominent repeat-unit monomers in the PHAs were 3-hydroxydodecanoate, 3-hydroxyoctanoate, 3-hydroxydodecanoate, and 3-hydroxytetradecenoate. This work represents the first description of fermentative mcl-PHA production from the soy molasses.
Sigma-Aldrich
DL-β-Hydroxylauric acid, ≥99% (GC)