Merck
  • Determination of phenols in water samples by single-drop microextraction followed by in-syringe derivatization and gas chromatography-mass spectrometric detection.
Determination of phenols in water samples by single-drop microextraction followed by in-syringe derivatization and gas chromatography-mass spectrometric detection.
Journal of chromatography. A (2005-11-30)
Mohammad Saraji, Mohsen Bakhshi
ABSTRACT
Trace analysis of phenolic compounds in water was performed by coupling single-drop microextraction (SDME) with in-syringe derivatization of the analytes and GC-MS analysis. The analytes were extracted from a 3ml sample solution using 2.5microl of hexyl acetate. After extraction, derivatization was carried out in syringe barrel using 0.5microl of N,O-bis(trimethylsilyl)acetamide. The influence of derivatizing reagent volume, derivatization time and temperature on the yield of the in-syringe silylation was investigated. Derivatization reaction is completed in 5min at 50 degrees C. Experimental SDME parameters, such as selection of organic solvent, sample pH, addition of salt, extraction time and temperature of extraction were studied. Analytical parameters, such as enrichment factor, precision, linearity and detection limits were also determined. The limits of detection were in the range of 4-61ng/l (S/N=3). The relative standard deviations obtained were between 4.8 and 12% (n=5).
Product Description
Supelco
N,O-Bis(trimethylsilyl)acetamide, for GC derivatization, LiChropur, ≥98.5% (GC)
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%