Merck
  • Prevalence and correction of 25(OH) vitamin D deficiency in peritoneal dialysis patients.
Prevalence and correction of 25(OH) vitamin D deficiency in peritoneal dialysis patients.
Peritoneal dialysis international : journal of the International Society for Peritoneal Dialysis (2005-07-19)
Nirav Shah, Judith Bernardini, Beth Piraino
ABSTRACT
Peritoneal dialysis (PD) patients are at risk for 25(OH) vitamin D deficiency due to effluent loss in addition to traditional risk factors. To measure 25(OH) vitamin D deficiency in prevalent PD patients, to evaluate a replacement dose, and to determine the effects of correction. 25(OH) vitamin D levels were drawn on prevalent PD patients. Patients deficientin 25(OH) vitamin D were given ergocalciferol, 50000 IU orally once per week for 4 weeks. Patients scored muscle weakness, bone pain, and fatigue on a scale of 0 (none) to 5 (severe). Serum calcium, phosphate, parathyroid hormone (PTH), and 25(OH) vitamin D, and 1,25(OH)2 vitamin D levels were obtained before and after treatment. 25(OH) vitamin D levels were measured in 29 PD patients. Deficiency (<15 ng/mL) was found in 28/29 (97%); 25/29 (86%) had undetectable levels (<7 ng/mL). One course of ergocalciferol corrected the deficiency in all but 1 patient, who required a second course. Scores for muscle weakness and bone pain fell from pre- to posttreatment (p < 0.001). 1,25(OH)2 vitamin D levels rose post ergocalciferol (from 20 to 26 pg/mL, n = 20, p = 0.09). Serum calcium, phosphate, and PTH levels did not change with ergocalciferol. Most PD patients had marked 25(OH) vitamin D deficiency, which was readily and safely corrected with one course of 50000 IU ergocalciferol, having no effect on serum calcium, phosphorus, or PTH, but complaints of muscle weakness and bone pain decreased. A prospective, placebo-controlled double-blinded study is needed to determine whether replacement of 25(PH) vitamin D is beneficial in PD patients.
Sigma-Aldrich
Ergocalciferol, 40,000,000 USP units/g
Ergocalciferol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ergocalciferol, ≥98.0% (sum of enantiomers, HPLC)
Supelco
Dihydrotachysterol, analytical standard
Supelco
Ergocalciferol (Vitamin D2), Pharmaceutical Secondary Standard; Certified Reference Material