Merck
  • Derivatization reaction of the mycotoxin moniliformin with 1,2-diamino-4,5-dichlorobenzene: structure elucidation of an unexpected reaction product by liquid chromatography/tandem mass spectrometry and liquid chromatography/nuclear magnetic resonance spectroscopy.
Derivatization reaction of the mycotoxin moniliformin with 1,2-diamino-4,5-dichlorobenzene: structure elucidation of an unexpected reaction product by liquid chromatography/tandem mass spectrometry and liquid chromatography/nuclear magnetic resonance spectroscopy.
Journal of mass spectrometry : JMS (2003-08-05)
Peter Zöllner, Annette Lienau, Klaus Albert, Wolfgang Lindner
ABSTRACT
The derivatization reaction of the mycotoxin moniliformin with 1,2-diamino-4,5-dichlorobenzene was previously introduced to improve distinctly the sensitivity of an assay applying high-performance liquid chromatography prior to fluorescence detection. In the course of the implementation of this derivatization approach into a liquid chromatographic/mass spectrometric method, an unexpected derivatization product has now been discovered by mass spectrometry. In order to elucidate its structure, detailed investigations with liquid chromatography/tandem mass spectrometry and liquid chromatography coupled on-line with NMR spectroscopy were performed. These studies give evidence for a heterocyclic structure that has been formed by the loss of one water and one carbon monoxide molecule. A reasonable mechanism for this derivatization reaction is proposed.
Sigma-Aldrich
4,5-Dichloro-o-phenylenediamine, 97%