Merck
  • ABC transporters and azole susceptibility in laboratory strains of the wheat pathogen Mycosphaerella graminicola.
ABC transporters and azole susceptibility in laboratory strains of the wheat pathogen Mycosphaerella graminicola.
Antimicrobial agents and chemotherapy (2002-11-19)
Lute-Harm Zwiers, Ioannis Stergiopoulos, Johannes G M Van Nistelrooy, Maarten A De Waard
ABSTRACT
Laboratory strains of Mycosphaerella graminicola with decreased susceptibilities to the azole antifungal agent cyproconazole showed a multidrug resistance phenotype by exhibiting cross-resistance to an unrelated chemical, cycloheximide or rhodamine 6G, or both. Decreased azole susceptibility was found to be associated with either decreased or increased levels of accumulation of cyproconazole. No specific relationship could be observed between azole susceptibility and the expression of ATP-binding cassette (ABC) transporter genes MgAtr1 to MgAtr5 and the sterol P450 14alpha-demethylase gene, CYP51. ABC transporter MgAtr1 was identified as a determinant in azole susceptibility since heterologous expression of the protein reduced the azole susceptibility of Saccharomyces cerevisiae and disruption of MgAtr1 in one specific M. graminicola laboratory strain with constitutive MgAtr1 overexpression restored the level of susceptibility to cyproconazole to wild-type levels. However, the level of accumulation in the mutant with an MgAtr1 disruption did not revert to the wild-type level. We propose that variations in azole susceptibility in laboratory strains of M. graminicola are mediated by multiple mechanisms.
Product Description
Supelco
Cyproconazol, PESTANAL®, analytical standard