Merck
  • Characterization of Na+-coupled glutamate/aspartate transport by a rat brain astrocyte line expressing GLAST and EAAC1.
Characterization of Na+-coupled glutamate/aspartate transport by a rat brain astrocyte line expressing GLAST and EAAC1.
The Journal of membrane biology (2001-06-27)
G A Kimmich, J Roussie, M Manglapus, J Randles
ABSTRACT
D-aspartate (D-Asp) uptake by suspensions of cerebral rat brain astrocytes (RBA) maintained in long-term culture was studied as a means of characterizing function and regulation of Glutamate/Aspartate (Glu/Asp) transporter isoforms in the cells. A-asp influx is Na+-dependent with Km = 5 microm and Vmax = 0.7 nmoles x min(-1) x mg protein-1. Influx is sigmoidal as f[Na+] with Na+Km approximately 12 microm and Hill coefficient of 1.9. The cells establish steady-state D-Asp gradients >3,000-fold. Phorbol ester (PMA) enhances uptake, and gradients near 6,000-fold are achieved due to a 2-fold increase in Vmax, with no change in Km. At initial [D-Asp] = 10 microm, RBA take up more than 90% of total D-Asp, and extracellular levels are reduced to levels below 1 microm. Ionophores that dissipate the Delta(mu)Na+ inhibit gradient formation. Genistein (GEN, 100 microm), a PTK inhibitor, causes a 40% decrease in d-Asp. Inactive analogs of PMA (4alpha-PMA) and GEN (daidzein) have no detectable effect, although the stimulatory PMA response still occurs when GEN is present. Further specificity of action is indicated by the fact that PMA has no effect on Na+-coupled ALA uptake, but GEN is stimulatory. d-Asp uptake is strongly inhibited by serine-O-sulfate (S-O-S), threohydroxy-aspartate (THA), L-Asp, and L-Glu, but not by D-Glu, kainic acid (KA), or dihydrokainate (DHK), an inhibition pattern characteristic of GLAST and EAAC1 transporter isoforms. mRNA for both isoforms was detected by RT-PCR, and Western blotting with appropriate antibodies shows that both proteins are expressed in these cells.