Merck
  • Synthesis and biological evaluation of ureido and thioureido derivatives of 2-amino-2-deoxy-D-glucose and related aminoalcohols as N-acetyl-beta-D-hexosaminidase inhibitors.
Synthesis and biological evaluation of ureido and thioureido derivatives of 2-amino-2-deoxy-D-glucose and related aminoalcohols as N-acetyl-beta-D-hexosaminidase inhibitors.
Carbohydrate research (1999-05-07)
C Tournaire-Arellano, S Y Hage, P Valès, R Caujolle, A Sanon, C Bories, P M Loiseau
ABSTRACT
Ureido and thioureido derivatives of 2-acetamido-2-deoxy-beta-D-glucose, 1-amino-1-deoxy-D-glucitol and 2-(2-aminoethoxy)ethanol were prepared as N-acetyl-beta-D-hexosaminidase (NAHase) inhibitors and were evaluated on Trichomonas vaginalis NAHase. Although none showed complete inhibition of the enzyme at 100 microM, 1-amino-1-deoxy-D-glucitol derivatives acted as competitive inhibitors of the NAHase of T. vaginalis.
Sigma-Aldrich
2-(2-Aminoethoxy)ethanol, 98%