Merck
  • Phenolic constituents of Amorpha fruticosa that inhibit NF-kappaB activation and related gene expression.
Phenolic constituents of Amorpha fruticosa that inhibit NF-kappaB activation and related gene expression.
Journal of natural products (2008-10-10)
Nguyen Tien Dat, Jeong-Hyung Lee, Kyeong Lee, Young-Soo Hong, Young Ho Kim, Jung Joon Lee
ABSTRACT
NF-kappaB is known to play a crucial role in the regulation of genes controlling the immune system, apoptosis, tumor cell growth, and tissue differentiation. Bioassay-guided fractionation of the n-hexane-soluble fraction of a methanol extract of Amorpha fruticosa afforded four new compounds, 5, 7, 8, and 9, and eight known compounds. Their structures were elucidated by spectroscopic methods. All compounds inhibited NF-kappaB activity, and tephrosin (1), 11-hydroxytephrosin (2), and deguelin (3) were the most active, with IC50 values of 0.11, 0.19, and 0.22 microM, respectively, in TNF-alpha-stimulated HeLa cell-based reporter gene assays. Further investigations showed that compounds 1, 5, and 6 blocked NF-kappaB/DNA binding activity and suppressed the expression of NF-kappaB target genes.
Sigma-Aldrich
Tephrosin, ≥95% (LC/MS-ELSD)
Sigma-Aldrich
Amorfrutin A, ≥90% (LC/MS-ELSD)