Merck
  • Amperometric bienzymatic biosensor for L-lactate analysis in wine and beer samples.
Amperometric bienzymatic biosensor for L-lactate analysis in wine and beer samples.
The Analyst (2012-07-06)
Sandra Pérez, Esteve Fàbregas
ABSTRACT
A novel amperometric bienzymatic biosensor has been developed based on the incorporation of Lactate Oxidase (LOx) and Horseradish Peroxidase (HRP) into a carbon nanotube/polysulfone membrane by the phase inversion technique onto screen-printed electrodes (SPEs). In order to improve the sensitivity and reduce the working potential, experimental conditions have been optimized and ferrocene has also been incorporated into the membrane as a redox mediator of the enzymatic reactions, which allows the reduction of H(2)O(2) at -100 mV. Measurements were carried out in phosphate buffer solution at pH 7.5 and under batch conditions. The biosensor response time to L-lactate was only 20 s and showed a good reproducibility (RSD 2.7%). Moreover, the detection limit was 0.05 mg L(-1) of l-lactate with a linear interval range from 0.1 mg L(-1) to 5 mg L(-1). Finally, the biosensor has been applied to the determination of l-lactic acid in different wine and beer samples. Then, the results obtained with the biosensor were compared with the ones obtained using, as a reference method, a commercial kit based on spectrophotometric measurements, obtaining an excellent agreement between the results, validating our approach.
Sigma-Aldrich
Polysulfone, average Mn ~22,000 by MO, beads
Sigma-Aldrich
Polysulfone, average Mw ~35,000 by LS, average Mn ~16,000 by MO, pellets (Transparent)