Merck
  • Glycoprotein labeling using engineered variants of galactose oxidase obtained by directed evolution.
Glycoprotein labeling using engineered variants of galactose oxidase obtained by directed evolution.
Journal of the American Chemical Society (2011-04-30)
Julie B Rannes, Avgousta Ioannou, Simon C Willies, Gideon Grogan, Carsten Behrens, Sabine L Flitsch, Nicholas J Turner
ABSTRACT
A directed evolution approach has been used for the generation of variants of galactose oxidase (GOase) that can selectively oxidize glycans on glycoproteins. The aldehyde function introduced on the glycans D-mannose (Man) and D-N-acetyl glucosamine (GlcNAc) by the enzyme variants could then be used to label the glycoproteins and also whole cells that display mannosides on their surface.
Sigma-Aldrich
Galactose Oxidase from Dactylium dendroides, lyophilized powder, ≥3,000 units/g solid
Sigma-Aldrich
Galactose Oxidase from Dactylium dendroides, ≥30 units/mg solid