Merck
  • Na,K-ATPase signal transduction triggers CREB activation and dendritic growth.
Na,K-ATPase signal transduction triggers CREB activation and dendritic growth.
Proceedings of the National Academy of Sciences of the United States of America (2009-01-24)
Luc Desfrere, Marie Karlsson, Hiromi Hiyoshi, Seth Malmersjö, Evanthia Nanou, Manuel Estrada, Ayako Miyakawa, Hugo Lagercrantz, Abdeljabbar El Manira, Mark Lal, Per Uhlén
ABSTRACT
Dendritic growth is pivotal in the neurogenesis of cortical neurons. The sodium pump, or Na,K-ATPase, is an evolutionarily conserved protein that, in addition to its central role in establishing the electrochemical gradient, has recently been reported to function as a receptor and signaling mediator. Although a large body of evidence points toward a dual function for the Na,K-ATPase, few biological implications of this signaling pathway have been described. Here we report that Na,K-ATPase signal transduction triggers dendritic growth as well as a transcriptional program dependent on cAMP response element binding protein (CREB) and cAMP response element (CRE)-mediated gene expression, primarily regulated via Ca(2+)/calmodulin-dependent protein (CaM) kinases. The signaling cascade mediating dendritic arbor growth also involves intracellular Ca(2+) oscillations and sustained phosphorylation of mitogen-activated protein (MAP) kinases. Thus, our results suggest a novel role for the Na,K-ATPase as a modulator of dendritic growth in developing neurons.
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder