Merck
  • PLK1-activating IFI16-STING-TBK1 pathway induces apoptosis of intestinal epithelial cells in patients with intestinal Behçet's syndrome.
PLK1-activating IFI16-STING-TBK1 pathway induces apoptosis of intestinal epithelial cells in patients with intestinal Behçet's syndrome.
The FEBS journal (2024-04-28)
Hua-Fang Bao, Chun-Hui She, Cheng-Cheng Hou, Da-Nian Ji, Dan Hu, Jun Zou, Yan Shen, Lei-Lei Jian, Jian-Fei Cai, Jing-Fen Ye, Dan Luo, Hai-Fen Ma, Jian-Long Guan
ABSTRACT
Inflammatory signals from immunological cells may cause damage to intestinal epithelial cells (IECs), resulting in intestinal inflammation and tissue impairment. Interferon-γ-inducible protein 16 (IFI16) was reported to be involved in the pathogenesis of Behçet's syndrome (BS). This study aimed to investigate how inflammatory cytokines released by immunological cells and IFI16 participate in the pathogenesis of intestinal BS. RNA sequencing and real-time quantitative PCR (qPCR) showed that the positive regulation of tumor necrosis factor-α (TNF-α) production in peripheral blood mononuclear cells (PBMCs) of intestinal BS patients may be related to the upregulation of polo like kinase 1 (PLK1) in PBMCs (P = 0.012). The plasma TNF-α protein level in intestinal BS was significantly higher than in healthy controls (HCs; P = 0.009). PBMCs of intestinal BS patients and HCs were co-cultured with human normal IECs (NCM460) to explore the interaction between immunological cells and IECs. Using IFI16 knockdown, PBMC-NCM460 co-culture, TNF-α neutralizing monoclonal antibody (mAb), stimulator of interferon genes (STING) agonist 2'3'-cGAMP, and the PLK1 inhibitor SBE 13 HCL, we found that PLK1 promotes the secretion of TNF-α from PBMCs of intestinal BS patients, which causes overexpression of IFI16 and induces apoptosis of IECs via the STING-TBK1 pathway. The expressions of IFI16, TNF-α, cleaved caspase 3, phosphorylated STING (pSTING) and phosphorylated tank binding kinase 1 (pTBK1) in the intestinal ulcer tissue of BS patients were significantly higher than that of HCs (all P < 0.05). PLK1 in PBMCs of intestinal BS patients increased TNF-α secretion, inducing IEC apoptosis via activation of the IFI16-STING-TBK1 pathway. PLK1 and the IFI16-STING-TBK1 pathway may be new therapeutic targets for intestinal BS.
Sigma-Aldrich
2′,3′-cGAMP sodium salt, ≥98% (HPLC)