Merck
  • Lack of canonical thyroid hormone receptor α signaling changes regulatory T cell phenotype in female mice.
Lack of canonical thyroid hormone receptor α signaling changes regulatory T cell phenotype in female mice.
iScience (2024-08-23)
Christina Wenzek, Devon Siemes, G Sebastian Hönes, Eva Pastille, Nina Härting, Frank Kaiser, Lars C Moeller, Daniel R Engel, Astrid M Westendorf, Dagmar Führer
ABSTRACT
The immune system has emerged as an important target of thyroid hormones (THs); however, the role of TH in T cells has so far remained elusive. In this study, we assessed the effect of TH receptor α (TRα) signaling on activation and function of T cells. Our findings show that lack of canonical TRα action not only increased the frequency of regulatory T cells (Treg) but propelled an activated and migratory Treg phenotype and nuclear factor κB (NF-κB) activation in Treg. Conversely, canonical TRα action reduced activation of the NF-κB pathway previously shown to play a pivotal role in Treg differentiation and function. Taken together, our findings demonstrate that TRα impacts T cell differentiation and phenotype. Given the well-known interaction of inflammation, immune responses, and TH axis in e.g., severe illness, altered TH-TRα signaling may have an important role in regulating T cell responses during disease.
Sigma-Aldrich
1(S),9(R)-(−)-Bicuculline methbromide, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-α-Tubulin Antibody, clone DM1A, clone DM1A, Upstate®, from mouse
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)