Merck
  • The intracellular interplay between galectin-1 and FGF12 in the assembly of ribosome biogenesis complex.
The intracellular interplay between galectin-1 and FGF12 in the assembly of ribosome biogenesis complex.
Cell communication and signaling : CCS (2024-03-12)
Aleksandra Gędaj, Aleksandra Chorążewska, Krzysztof Ciura, Radosław Karelus, Dominika Żukowska, Martyna Biaduń, Marta Kalka, Małgorzata Zakrzewska, Natalia Porębska, Łukasz Opaliński
ABSTRACT
Galectins constitute a class of lectins that specifically interact with β-galactoside sugars in glycoconjugates and are implicated in diverse cellular processes, including transport, autophagy or signaling. Since most of the activity of galectins depends on their ability to bind sugar chains, galectins exert their functions mainly in the extracellular space or at the cell surface, which are microenvironments highly enriched in glycoconjugates. Galectins are also abundant inside cells, but their specific intracellular functions are largely unknown. Here we report that galectin-1, -3, -7 and -8 directly interact with the proteinaceous core of fibroblast growth factor 12 (FGF12) in the cytosol and in nucleus. We demonstrate that binding of galectin-1 to FGF12 in the cytosol blocks FGF12 secretion. Furthermore, we show that intracellular galectin-1 affects the assembly of FGF12-containing nuclear/nucleolar ribosome biogenesis complexes consisting of NOLC1 and TCOF1. Our data provide a new link between galectins and FGF proteins, revealing an unexpected glycosylation-independent intracellular interplay between these groups of proteins.