- Controlled chemical modification of hyaluronic acid: synthesis, applications, and biodegradation of hydrazide derivatives.
Controlled chemical modification of hyaluronic acid: synthesis, applications, and biodegradation of hydrazide derivatives.
Journal of controlled release : official journal of the Controlled Release Society (1998-09-19)
G D Prestwich, D M Marecak, J F Marecek, K P Vercruysse, M R Ziebell
PMID9741917
ABSTRACT
Controlled modification of the carboxylic acid moieties of hyaluronic acid with mono- and polyfunctional hydrazides leads to biochemical probes, biopolymers with altered physical and chemical properties, tethered drugs for controlled release, and crosslinked hydrogels as biocompatible scaffoldings for tissue engineering. Methods for polyhydrazide synthesis, for prodrug preparation, for hydrogel crosslinking, and for monitoring biodegradation are described.