Merck
Electroporation-Mediated CRISPR/Cas9 Genome Editing in Rat Zygotes.
Methods in molecular biology (Clifton, N.J.) (2023-03-31)
Daniel J Davis, Hongsheng Men, Elizabeth C Bryda
ABSTRACT
Genetic engineering in the rat has been revolutionized by the development of CRISPR-based genome editing tools. Conventional methods for inserting genome editing elements such as CRISPR/Cas9 reagents into rat zygotes include cytoplasmic or pronuclear microinjections. These techniques are labor-intensive, require specialized micromanipulator equipment, and are technically challenging. Here, we describe a simple and effective method for zygote electroporation in which CRISPR/Cas9 reagents are introduced into rat zygotes via pores produced by precise electrical pulses applied to the cells. Zygote electroporation allows for high-throughput efficient genome editing in rat embryos.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)