Merck
Automated multi-target super-resolution microscopy with trust regions.
Biomedical optics express (2023-01-27)
David J Schodt, Farzin Farzam, Sheng Liu, Keith A Lidke
ABSTRACT
We describe a dedicated microscope for automated sequential localization microscopy which we term Sequential Super-resolution Microscope (SeqSRM). This microscope automates precise stage stabilization on the order of 5-10 nanometers and data acquisition of all user-selected cells on a coverslip, limiting user interaction to only cell selection and buffer exchanges during sequential relabeling. We additionally demonstrate that nanometer-scale changes to cell morphology affect the fidelity of the resulting multi-target super-resolution overlay reconstructions generated by sequential super-resolution microscopy, and that regions affected by these shifts can be reliably detected and masked out using brightfield images collected periodically throughout the experiment. The SeqSRM enables automated multi-target imaging on multiple user-selected cells without the need for multiple distinct fluorophores and emission channels, while ensuring that the resulting multi-target localization data accurately reflect the relative organization of the underlying targets.
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture