Merck
  • Image-guided lymph node core-needle biopsy predicts survival in mycosis fungoides and Sézary syndrome.
Image-guided lymph node core-needle biopsy predicts survival in mycosis fungoides and Sézary syndrome.
The British journal of dermatology (2021-01-06)
J Calvani, A de Masson, C de Margerie-Mellon, É de Kerviler, C Ram-Wolff, A Gruber, V Meignin, P Brice, A Sadoux, S Mourah, M Bagot, M Battistella
ABSTRACT
The prognosis of Sézary syndrome (SS) and mycosis fungoides (MF) depends on lymph node (LN) involvement. The usefulness of LN image-guided core-needle biopsies (CNBs), instead of surgical sampling, has been poorly evaluated. To determine the prognostic value of LN CNB in MF/SS. A retrospective search was conducted to identify all LN biopsy specimens of MF/SS between 2008 and 2019. Biopsies were staged according to the International Society for Cutaneous Lymphomas/European Organisation for Research and Treatment of Cancer (ISCL/EORTC) criteria. We performed immunolabelling and determined the tumour clone frequency (TCF) by high-throughput sequencing of the T-cell receptor beta locus. We included 119 consecutive biopsies from 100 patients, 45 with MF and 55 with SS. N1, N2 and N3 stages were diagnosed in 34 (29%), 26 (22%) and 59 (49%) cases, respectively. The TCF, Ki67 index, and percentage of cells positive for thymocyte selection-associated high mobility group box protein (TOX), programmed cell death protein 1 (PD1), killer cell immunoglobulin-like receptor 3DL2 (KIR3DL2) and cluster of differentiation (CD)30 were all positively correlated with the N stage. Median overall survival (OS) for N1/N2 vs. N3 patients was 42 months (range 26-not reached) vs. 14 months (range 5-30), respectively (P < 0·001). In univariate analyses, an age > 75 years, LN short-axis diameter > 15 mm, N3 stage, presence of large-cell transformation, TOX > 60%, PD1 > 25%, Ki67 > 30%, KIR3DL2 > 15%, CD30 > 10% and TCF > 25% were identified as adverse prognostic factors. In multivariate analyses, only an age > 75 years and Ki67 index > 30% were associated with reduced OS. We developed a new prognostic index associating the N stage and the Ki67 index, which better discriminates N3 patients with poor prognosis. CNB allows an objective assessment of the LN involvement in MF/SS, relevant for staging and prognosis.