Merck
  • Attenuated kallikrein-related peptidase activity disrupts desquamation and leads to stratum corneum thickening in human skin equivalent models.
Attenuated kallikrein-related peptidase activity disrupts desquamation and leads to stratum corneum thickening in human skin equivalent models.
The British journal of dermatology (2016-07-22)
J A McGovern, C Meinert, S J de Veer, B G Hollier, T J Parker, Z Upton
ABSTRACT
Epidermal homeostasis is maintained through the balance between keratinocyte proliferation, differentiation and desquamation; however, human skin equivalent (HSE) models are known to differentiate excessively. In native tissue, proteases such as kallikrein-related peptidase (KLK) 5 and KLK7 cleave the extracellular components of corneodesmosomes; proteins corneodesmosin, desmocollin 1 and desmoglein 1, loosening the cellular connections and enabling desquamation. The actions of KLK7 are tightly controlled by protease inhibitors, skin-derived antileucoproteinase (SKALP) and lymphoepithelial Kazal-type-related inhibitor (LEKTI), which also inhibits KLK5, localizing protease activity to the stratum corneum. To investigate the mechanisms that inhibit the desquamation cascade in HSE models. Human skin tissue and HSE models were investigated using gene microarray, real-time polymerase chain reaction (PCR), immunohistochemistry and Western blot analysis to examine key components of the desquamation pathway. To elucidate proteolytic activity in HSEs and native skin, in situ and gel zymography was performed. Histological analysis indicated that HSE models form a well-organized epidermis, yet develop an excessively thick and compact stratum corneum. Gene microarray analysis revealed that the desquamation cascade was dysregulated in HSE models and this was confirmed using real-time PCR and immunohistochemistry. Immunohistochemistry and Western blot indicated overexpression of LEKTI and SKALP in HSEs. Although KLK7 was also highly expressed in HSEs, zymography indicated that protease activation and activity was lower than in native skin. These findings demonstrate that stratum corneum thickening is due to inhibited KLK5 and KLK7 activation and a subsequent lack of corneodesmosome degradation in the HSE model epidermis.