Merck
  • Overexpression of regucalcin blocks the migration, invasion, and bone metastatic activity of human prostate cancer cells: Crosstalk between cancer cells and bone cells.
Overexpression of regucalcin blocks the migration, invasion, and bone metastatic activity of human prostate cancer cells: Crosstalk between cancer cells and bone cells.
The Prostate (2022-04-03)
Masayoshi Yamaguchi, Tomiyasu Murata, Joe W Ramos
ABSTRACT
Prostate cancer is a bone metastatic cancer and is the second leading cause of cancer-related death in men. Prolonged progression-free survival of prostate cancer patients is associated with high regucalcin expression in the tumor tissues. This study investigates the underlying mechanism by which regucalcin prevents bone metastatic activity of prostate cancer cells. Human prostate cancer PC-3 or DU-145 wild-type cells or regucalcin-overexpressing PC-3 or DU-145 cells (transfectants) were cultured in Dulbecco's modified Eagle's medium containing 10% fetal bovine serum. Overexpressed regucalcin suppressed the migration and invasion of bone metastatic human prostate cancer cells in vitro, and it reduced the levels of key proteins in metastasis including Ras, Akt, MAPK, RSK-2, mTOR, caveolin-1, and integrin β1. Invasion of prostate cancer cells was promoted by coculturing with preosteoblastic MC3T3-E1 or preosteoclastic RAW264.7 cells. Coculturing with cancer cells and bone cells repressed the growth of preosteoblastic cells and enhanced osteoclastogenesis of preosteoclastic cells, and these alterations were caused by a conditioned medium from cancer cell culture. Disordered differentiation of bone cells was prevented by regucalcin overexpression. Production of tumor necrosis factor-α (TNF-α) in cancer cells was blocked by overexpressed regucalcin. Of note, the effects of conditioned medium on bone cells were prevented by NF-κB inhibitor. TNF-α may be important as a mediator in the crosstalk between cancer cells and bone cells. Overexpression of regucalcin suppressed the migration, invasion, and bone metastatic activity of human prostate cancer cells. This study may provide a new strategy for therapy with the regucalcin gene transfer.