Merck
  • Delete and repeat: a comprehensive toolkit for sequential gene knockout in the budding yeast Saccharomyces cerevisiae.
Delete and repeat: a comprehensive toolkit for sequential gene knockout in the budding yeast Saccharomyces cerevisiae.
Methods in molecular biology (Clifton, N.J.) (2011-08-05)
Johannes H Hegemann, Sven Boris Heick
ABSTRACT
Gene inactivation is an essential step in the molecular dissection of gene function. In the yeast Saccharomyces cerevisiae, many tools for gene disruption are available. Gene disruption cassettes comprising completely heterologous marker genes flanked by short DNA segments homologous to the regions to the left and right of the gene to be deleted mediate highly efficient one-step gene disruption events. Routinely, in more than 50% of analyzed clones, the marker cassette is integrated in the targeted location. The inclusion of loxP sites flanking the disruption marker gene allows sequence-specific Cre recombinase-mediated marker rescue so that the marker can be reused to disrupt another gene. Here, we describe a comprehensive toolbox for multiple gene disruptions comprising a set of seven heterologous marker genes including four dominant resistance markers for gene disruption, plus a set of Cre expression plasmids carrying eight different selection markers, four of them dominant.
Product Description
Millipore
G 418 Sulfate, Cell Culture Tested, G418 also known as Geneticin is an aminoglycoside antibiotic related to Gentamicin. Used as a selective agent in transfection of eukaryotic cells. Has highest potency ≥730 µg/mg and purity ≥98%.