Merck
  • An Optimized Stress Granule Detection Method: Investigation of UBQLN2 Effect on Stress Granule Formation.
An Optimized Stress Granule Detection Method: Investigation of UBQLN2 Effect on Stress Granule Formation.
Methods in molecular biology (Clifton, N.J.) (2022-11-01)
Elizabeth J Alexander, Jiou Wang
ABSTRACT
Stress granules (SGs) are cytosolic ribonucleoprotein granules that form via a liquid-liquid phase separation in response to environmental stresses such as heat, oxidative, and osmotic changes. Due to the condensation of low complexity, hydrophobic regions in core SG components in these highly dynamic granules, defects in SG maintenance and formation have been linked to toxic aggregate formation in neurodegenerative diseases such as amyotrophic lateral sclerosis and frontotemporal dementia. However, efforts to dissect mechanisms regulating SG formation and maintenance have been limited by methods of tracking protein-SG localization. Here we describe a method for detecting and quantifying recruitment of cytosolically enriched proteins to SGs by indirect immunofluorescence microscopy. Using this method, we tracked the transient recruitment of the cytosolically enriched ubiquitin-like protein, ubiquilin 2 (UBQLN2), and a number of other factors into SGs, demonstrating its utility (Alexander et al., Proc Natl Acad Sci U S A 115:E11485-E11494, 2018).