Merck
  • Cell shape anisotropy contributes to self-organized feather pattern fidelity in birds.
Cell shape anisotropy contributes to self-organized feather pattern fidelity in birds.
PLoS biology (2022-10-11)
Camille Curantz, Richard Bailleul, María Castro-Scherianz, Magdalena Hidalgo, Melina Durande, François Graner, Marie Manceau
ABSTRACT
Developing tissues can self-organize into a variety of patterned structures through the stabilization of stochastic fluctuations in their molecular and cellular properties. While molecular factors and cell dynamics contributing to self-organization have been identified in vivo, events channeling self-organized systems such that they achieve stable pattern outcomes remain unknown. Here, we described natural variation in the fidelity of self-organized arrays formed by feather follicle precursors in bird embryos. By surveying skin cells prior to and during tissue self-organization and performing species-specific ex vivo drug treatments and mechanical stress tests, we demonstrated that pattern fidelity depends on the initial amplitude of cell anisotropy in regions of the developing dermis competent to produce a pattern. Using live imaging, we showed that cell shape anisotropy is associated with a limited increase in cell motility for sharp and precisely located primordia formation, and thus, proper pattern geometry. These results evidence a mechanism through which initial tissue properties ensure stability in self-organization and thus, reproducible pattern production.